SY3H3D678660 Synthonix Corporation

5-(2-Bromoacetyl)-2-hydroxybenzaldehyde

  • MDL number MFCD09833486
  • Cas Number : 115787-50-3
  • Molecular Weight : 243.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/51280.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/51280.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart