SY3H3D678674 Synthonix Corporation

2-(4-Bromophenyl)propanenitrile

  • MDL number MFCD11036513
  • Cas Number : 42186-06-1
  • Molecular Weight : 210.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/51451.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/51451.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart