SY3H3D678675 Synthonix Corporation

3-(4-Bromophenyl)propanenitrile

  • MDL number MFCD01310795
  • Cas Number : 57775-08-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/51452.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/51452.html
pictograms GHS07
hstatements H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart