SY3H3D678677 Synthonix Corporation

7-Bromo-6-methoxyisoindolin-1-one

  • MDL number MFCD21605666
  • Cas Number : 1226879-79-3
  • Molecular Weight : 242.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/51469.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/51469.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart