SY3H3D678687 Synthonix Corporation

2-(5-Bromo-2-methoxyphenyl)acetonitrile

  • MDL number MFCD00016392
  • Cas Number : 7062-40-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form Pale-yellow to Yellow to dark-yellow powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/51518.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/51518.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart