SY3H3D678693 Synthonix Corporation

3-BROMO-5-FLUORO-2-METHYL-BENZOIC ACID METHYL ESTER

  • MDL number No MDL found
  • Cas Number : 1187318-53-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/51592.html

Safety Information

MSDS https://synthonix.com/51592.html
Availability details for
Quantity
  • Availability
  • Add to Cart