SY3H3D6786A4 Synthonix Corporation

3-Bromo-4-ethoxybenzaldehyde

  • MDL number MFCD00672941
  • Cas Number : 108373-05-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Crystal - Powder / Colour: White - Slightly pale yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/51877.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/51877.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart