SY3H3D6786AA Synthonix Corporation

4-(Bromomethyl)phenyl acetate

  • MDL number No MDL found
  • Cas Number : 52727-95-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/51883.html

Safety Information

MSDS https://synthonix.com/51883.html
Availability details for
Quantity
  • Availability
  • Add to Cart