SY3H3D6786BC Synthonix Corporation

4-Bromo-2,5-dimethoxybenzaldehyde

  • MDL number MFCD00156964
  • Cas Number : 31558-41-5
  • Molecular Weight : 245.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/51974.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/51974.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart