SY3H3D6786C4 Synthonix Corporation

6-Bromochroman

  • MDL number MFCD10698725
  • Cas Number : 3875-78-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. refrigerator
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/52016.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/52016.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart