SY3H3D6786DC Synthonix Corporation

(R)-tert-Butyl pyrrolidine-2-carboxylate hydrochloride

  • MDL number MFCD00153460
  • Cas Number : 184719-80-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Crystal - Powder / Colour: White - Red
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/52206.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/52206.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart