SY3H3D6786DD Synthonix Corporation

(S)-1-tert-Butyl 4-methyl 2-aminosuccinate hydrochloride

  • MDL number MFCD05662354
  • Cas Number : 34582-30-4
  • Molecular Weight : 239.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/52209.html

Safety Information

signalword Warning
pcodes P261,P305,P351,P338
MSDS https://synthonix.com/52209.html
pictograms GHS07
hstatements H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart