SY3H3D6786E0 Synthonix Corporation

(R)-tert-Butyl 3-aminopyrrolidine-1-carboxylate hydrochloride

  • MDL number MFCD08061978
  • Cas Number : 1004538-34-4
  • Molecular Weight : 222.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/52212.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/52212.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart