SY3H3D678701 Synthonix Corporation

5-(4-Bromophenyl)pyrrolidin-2-one

  • MDL number MFCD15527558
  • Cas Number : 207989-90-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/52394.html

Safety Information

signalword Warning
pcodes P280
MSDS https://synthonix.com/52394.html
pictograms GHS07
hstatements H302, H317
Availability details for
Quantity
  • Availability
  • Add to Cart