SY3H3D678703 Synthonix Corporation

3-Bromo-4,5-dihydro-1H-benzo[b]azepin-2(3H)-one

  • MDL number MFCD03085918
  • Cas Number : 86499-96-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Crystal - Powder / Colour: White - Yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/52396.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/52396.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart