SY3H3D678707 Synthonix Corporation

2-Bromo-1-(4-chlorophenyl)butan-1-one

  • MDL number MFCD12031566
  • Cas Number : 1011-26-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/52495.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/52495.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart