SY3H3D67871C Synthonix Corporation

2-(4-(Bromomethyl)phenyl)propanoic acid

  • MDL number MFCD02093445
  • Cas Number : 111128-12-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Crystal - Powder
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/52631.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/52631.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart