SY3H3D678729 Synthonix Corporation

1-(5-Bromo-1H-pyrrolo[2,3-b]pyridin-3-yl)-N,N-dimethylmethanamine

  • MDL number MFCD13193319
  • Cas Number : 183208-54-0
  • Molecular Weight : 254.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/52711.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/52711.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart