SY3H3D67872D Synthonix Corporation

N-(4-Bromophenyl)pyrrolidine

  • MDL number MFCD04112480
  • Cas Number : 22090-26-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. refrigerator
Physical Form Crystal - Powder / Colour: White - Yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/52721.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/52721.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart