SY3H3D678730 Synthonix Corporation

(4-Bromophenyl)(cyclopropyl)methanamine

  • MDL number MFCD05192037
  • Cas Number : 90868-92-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/52724.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/52724.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart