SY3H3D678731 Synthonix Corporation

5-Bromo-1,2,3,4-tetrahydronaphthalen-1-amine

  • MDL number MFCD07374008
  • Cas Number : 405142-63-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/52725.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/52725.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart