SY3H3D678732 Synthonix Corporation

6-Bromo-1,2,3,4-tetrahydronaphthalen-2-amine

  • MDL number MFCD06739165
  • Cas Number : 167355-41-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/52726.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/52726.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart