SY3H3D678734 Synthonix Corporation

7-Bromo-1,2,3,4-tetrahydronaphthalen-2-amine

  • MDL number MFCD06739164
  • Cas Number : 885280-71-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. refrigerator
Physical Form Pale-yellow or Pale-gray to Brown Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/52728.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/52728.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart