SY3H3D678737 Synthonix Corporation

2-(3-Bromophenyl)pyrrolidine hydrochloride

  • MDL number MFCD08456887
  • Cas Number : 1171898-22-8
  • Molecular Weight : 262.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form White to off-white solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/52731.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/52731.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart