SY3H3D678740 Synthonix Corporation

(2-((tert-Butoxycarbonyl)amino)pyrimidin-5-yl)boronic acid

  • MDL number MFCD16251533
  • Cas Number : 883231-25-2
  • Molecular Weight : 239.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/50423.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/50423.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart