SY3H3D678748 Synthonix Corporation

4-(2-Bromophenyl)morpholine

  • MDL number MFCD03086182
  • Cas Number : 87698-82-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/52768.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/52768.html
pictograms GHS07
hstatements H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart