SY3H3D678749 Synthonix Corporation

4-(3-Bromophenyl)morpholine

  • MDL number MFCD04112585
  • Cas Number : 197846-82-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Referigrator
Physical Form Light-yellow to yellow liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/52769.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/52769.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart