SY3H3D678758 Synthonix Corporation

1-Bromo-3-(tert-butyl)benzene

  • MDL number MFCD00234661
  • Cas Number : 3972-64-3
  • Molecular Weight : 213.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/52872.html

Safety Information

signalword Warning
pcodes P261,P305,P351,P338
MSDS https://synthonix.com/52872.html
pictograms GHS07
hstatements H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart