SY3H3D67875D Synthonix Corporation

3-Boc-Amino-4-bromopyridine

  • MDL number MFCD05664029
  • Cas Number : 885275-14-9
  • Molecular Weight : 273.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/52886.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/52886.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart