SY3H3D678767 Synthonix Corporation

N-(tert-Butyl)-6-chloronicotinamide

  • MDL number MFCD08460992
  • Cas Number : 115309-58-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form White to off-white solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/52900.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/52900.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart