SY3H3D67876C Synthonix Corporation

tert-Butyl isonicotinate

  • MDL number MFCD19600069
  • Cas Number : 81660-73-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form Yellow Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/52938.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/52938.html
pictograms GHS07
hstatements H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart