SY3H3D678774 Synthonix Corporation

2-Bromo-4-tert-butylphenol

  • MDL number MFCD02682891
  • Cas Number : 2198-66-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Liquid - Crystal - Lump
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/53013.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/53013.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart