SY3H3D678778 Synthonix Corporation

2-(3-Bromophenyl)-2-methylpropan-1-ol

  • MDL number MFCD11040622
  • Cas Number : 81606-48-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/53017.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/53017.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart