SY3H3D678789 Synthonix Corporation

2-(Benzyloxy)propan-1-ol

  • MDL number MFCD24448608
  • Cas Number : 70448-03-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Pale-yellow to Yellow-brown Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/53085.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/53085.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart