SY3H3D67878F Synthonix Corporation

2-(sec-Butyl)aniline

  • MDL number MFCD00040573
  • Cas Number : 55751-54-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. refrigerator
Physical Form Brown Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/53112.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/53112.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart