SY3H3D678792 Synthonix Corporation

N-Boc-3-Amino-4-iodopyridine

  • MDL number MFCD04973410
  • Cas Number : 154048-89-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/53121.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/53121.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart