SY3H3D678795 Synthonix Corporation

tert-Butyl 6-aminopyridin-2-ylcarbamate

  • MDL number MFCD09037865
  • Cas Number : 322690-31-3
  • Molecular Weight : 209.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Crystal - Powder / Colour: White - Slightly pale yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/53129.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/53129.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart