SY3H3D67879D Synthonix Corporation

tert-Butyl (5-methylthiophen-3-yl)carbamate

  • MDL number No MDL found
  • Cas Number : 1251734-12-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/53157.html

Safety Information

MSDS https://synthonix.com/53157.html
Availability details for
Quantity
  • Availability
  • Add to Cart