SY3H3D6787A9 Synthonix Corporation

4-(tert-Butyl)benzene-1,2-diamine

  • MDL number MFCD00052695
  • Cas Number : 68176-57-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Yellow to Brown to Black Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/53179.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/53179.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart