SY3H3D6787B0 Synthonix Corporation

tert-Butyl 3-cyano-3-hydroxypyrrolidine-1-carboxylate

  • MDL number MFCD11878020
  • Cas Number : 1194376-31-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form White to off-white powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/53191.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/53191.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart