SY3H3D6787B5 Synthonix Corporation

tert-Butyl 5,6-dihydro-[1,2,4]triazolo[4,3-a]pyrazine-7(8H)-carboxylate

  • MDL number MFCD10574958
  • Cas Number : 723286-79-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/53202.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/53202.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart