SY3H3D6787BC Synthonix Corporation

tert-Butyl 3-acetoxyazetidine-1-carboxylate

  • MDL number MFCD24445131
  • Cas Number : 1215205-53-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Yellow to light-brown liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/53236.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/53236.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart