SY3H3D6787D9 Synthonix Corporation

1-Boc-3-Carbamoylpiperazine

  • MDL number MFCD09608050
  • Cas Number : 112257-24-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. refrigerator
Physical Form White to off-white powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/53286.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/53286.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart