SY3H3D6787DE Synthonix Corporation

(2R,4S)-tert-Butyl 4-hydroxy-2-methylpyrrolidine-1-carboxylate

  • MDL number MFCD20278359
  • Cas Number : 348165-62-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/53296.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/53296.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart