SY3H3D6787F9 Synthonix Corporation

1-Bromo-3,7-dimethyloctane

  • MDL number MFCD00039201
  • Cas Number : 3383-83-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Clear liquid / Colour: Colorless - Yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/53343.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/53343.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart