SY3H3D678801 Synthonix Corporation

2-(tert-Butyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

  • MDL number MFCD06657801
  • Cas Number : 99810-76-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/53357.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/53357.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart