SY3H3D678805 Synthonix Corporation

1-((tert-Butyldimethylsilyl)oxy)butan-2-amine

  • MDL number MFCD03001670
  • Cas Number : 811841-81-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. refrigerator
Physical Form Colorless to light-yellow liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/53371.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/53371.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart