SY3H3D678806 Synthonix Corporation

1,3-Bis(aminopropyl)tetramethyldisiloxane

  • MDL number No MDL found
  • Cas Number : 2469-55-8
  • Molecular Weight : 248.52

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 83
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/53372.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/53372.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart