SY3H3D678836 Synthonix Corporation

1-(2,4-Bis(trifluoromethyl)phenyl)ethanone

  • MDL number MFCD00042495
  • Cas Number : 237069-82-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Pale-yellow to Yellow to Brown Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/53498.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/53498.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart