SY3H3D678837 Synthonix Corporation

2-Bromo-5-phenylthiophene

  • MDL number MFCD00159575
  • Cas Number : 29488-24-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Crystal - Powder / Colour: White - Slightly pale yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/53499.html

Safety Information

signalword Warning
pcodes P280
MSDS https://synthonix.com/53499.html
pictograms GHS07
hstatements H317, H319
Availability details for
Quantity
  • Availability
  • Add to Cart